Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Antibody (OX-1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10064895K Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064895K 0.05 mg
NB10064895 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064895K Supplier Novus Biologicals Supplier No. NB10064895SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 18 publications

CD45 Monoclonal specifically detects CD45 in Caenorhabditis elegans, Human, Rat samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen CD45
Applications CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OX-1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P04157
Gene Alias B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Gene Symbols PTPRC
Host Species Mouse
Immunogen A 100kD thymocyte glycoprotein fraction enriched for the rat leukocyte common antigen (L-CA).
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Hematopoietic Stem Cell Markers, Immunology, Immunophenotyping, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5788
Target Species C. elegans, Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.