Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45 Antibody (YKIX716.13), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB10065906J Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065906J 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065906J Supplier Novus Biologicals Supplier No. NB10065906APC
Add to Cart
Add to Cart

Rat Monoclonal Antibody has been used in 3 publications

CD45 Monoclonal specifically detects CD45 in Canine, Equine samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen CD45
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone YKIX716.13
Conjugate APC
Dilution Flow Cytometry
Gene Alias B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Gene Symbols PTPRC
Host Species Rat
Immunogen Canine thymocytes
Molecular Weight of Antigen 147 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5788
Test Specificity Recognizes canine CD45 also known as leukocyte common antigen lustered as Canine CD45 in the First Canine Leukocyte Antigen Workshop. CD45 is expressed on all leukocytes in canine peripheral blood and may be against CD45RB isoform. This antibody reacts with CD45 on all outbred mongrels and beagles tested.
Target Species Canine, Equine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.