Learn More
CD45 Mouse anti-Human, APC-Cy7, Clone: 2D1, BD
Mouse Monoclonal Antibody
$164.00 - $373.50
Specifications
Antigen | CD45 |
---|---|
Clone | 2D1 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC-Cyanine7 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561863
|
BD Biosciences
561863 |
25 Tests |
Each for $164.00
|
|
|||||
BDB557833
|
BD Biosciences
557833 |
100 Tests |
Each for $373.50
|
|
|||||
Description
The 2D1 monoclonal antibody recognizes an epitope on all forms of CD45, a tyrosine phosphatase. CD45 is a type I transmembrane glycoprotein that belongs to the Protein Tyrosine Phosphatase (PTP) family. There are several isoforms of CD45 with molecular weights ranging from 180 to 220 kDa. CD45 is expressed on all nucleated cells of hematopoietic origin and is also known as the Leukocyte Common Antigen (LCA). CD45 is most highly expressed on lymphocytes and multiple CD45 isoforms can be coexpressed on individual leucocytes. CD45 plays an important role in regulating leucocyte functions by modifying other signaling molecules.
Specifications
CD45 | |
Flow Cytometry | |
APC-Cyanine7 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
2D1 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified | |
PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.