Learn More
CD45 Mouse anti-Rat, FITC, Clone: OX-1, BD
Mouse Monoclonal Antibody
$116.50 - $404.00
Specifications
Antigen | CD45 |
---|---|
Clone | OX-1 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561867
|
BD Biosciences
561867 |
50 μg |
Each for $116.50
|
|
|||||
BDB554877
|
BD Biosciences
554877 |
0.5 mg |
Each for $404.00
|
|
|||||
Description
The OX-1 antibody reacts with all molecular forms of CD45 (Leukocyte Common Antigen) on all hematopoietic cells except erythrocytes. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD45 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Rat | |
Leukocyte Common Antigen-enriched Glycoprotein Fraction from Wistar Rat Thymocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
OX-1 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
Ptprc; Lca; Leucocyte common antigen; RT7; T200 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.