Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RA Antibody (CA4.1D3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064897
Description
CD45RA Monoclonal specifically detects CD45RA in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD45RA | |
Monoclonal | |
Unconjugated | |
Tissue culture supernatant with 0.09% Sodium Azide | |
PTPRC | |
Made to Dog CD45RA | |
1 mL | |
Cell Biology, Cytokine Research, Hematopoietic Stem Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Stem Cell Markers | |
5788 | |
Canine | |
Supernatant |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
CA4.1D3 | |
Flow Cytometry Neat, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Mouse | |
Tissue culture supernatant | |
RUO | |
Primary | |
Recognizes the canine homologue of CD45RA, a cell surface glycoprotein expressed by B lymphocytes and a subset of T-lymphocytes. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction