Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RA Antibody (CA4.1D3), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | CD45RA |
---|---|
Clone | CA4.1D3 |
Dilution | Flow Cytometry Neat, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017660
![]() |
Novus Biologicals
NB100648970.25ML |
0.25 mL |
Each for $206.50
|
|
|||||
NB10064897
![]() |
Novus Biologicals
NB10064897 |
1 mL |
Each for $499.50
|
|
|||||
Description
CD45RA Monoclonal specifically detects CD45RA in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD45RA | |
Flow Cytometry Neat, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Supernatant | |
RUO | |
Canine | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
PTPRC | |
IgG1 | |
Tissue culture supernatant | |
Recognizes the canine homologue of CD45RA, a cell surface glycoprotein expressed by B lymphocytes and a subset of T-lymphocytes. |
CA4.1D3 | |
Immunohistochemistry, Immunohistochemistry (Frozen), Flow Cytometry | |
Unconjugated | |
Mouse | |
Cell Biology, Cytokine Research, Hematopoietic Stem Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Stem Cell Markers | |
Tissue culture supernatant with 0.09% Sodium Azide | |
5788 | |
Made to Dog CD45RA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title