Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RC Antibody (C455.1F), Biotin, Novus Biologicals™
SDP

Catalog No. NBP128134 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128134 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128134 Supplier Novus Biologicals Supplier No. NBP128134
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD45RC Monoclonal specifically detects CD45RC in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen CD45RC
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone C455.1F
Concentration 0.5mg/mL
Conjugate Biotin
Dilution Flow Cytometry 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500
Gene Alias B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Gene Symbols PTPRC
Host Species Rat
Immunogen An exon C-dependent epitope of CD45 glycoprotein
Purification Method Protein A or G purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5788
Test Specificity An exon C-dependent epitope of CD45 glycoprotein
Target Species Mouse
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.