Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RC Antibody (OX22), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB1206406R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1206406R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1206406R Supplier Novus Biologicals Supplier No. NB1206406R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD45RC Monoclonal specifically detects CD45RC in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CD45RC
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OX22
Conjugate DyLight 550
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Gene Symbols PTPRC
Host Species Mouse
Immunogen Rat lymphocytes stimulated in culture with phytohaemagglutinin.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5788
Test Specificity NB120-6406 recognizes rat CD45RC, the high molecular weight form of the leucocyte common antigen. This antigen is found on B cells, approximately 50% of bone marrow cells, all CD8+ve T cells, but splits CD4+ve T cells into two populations,CD4+ CD45RC (Th 1-like) and CD4+ CD45RC low (Th-2 like). This product is routinely tested in flow cytometry on rat splenocytes.
Target Species Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.