Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RO Antibody (UCHL1), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB297533 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297533 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297533 Supplier Novus Biologicals Supplier No. FAB10642Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CD45RO
Applications Flow Cytometry
Classification Monoclonal
Clone UCHL1
Conjugate DyLight 680
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Host Species Mouse
Immunogen Cultured T cells from an IL-2-dependent T-cell line (CA1) prepared from human peripheral blood activated with influenza virus.
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5788
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.