Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD46 Antibody (344519), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB366764 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB366764 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB366764 Supplier Novus Biologicals Supplier No. FAB2005D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD46 Monoclonal specifically detects CD46 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen CD46
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 344519
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD46, Cys35-Asp328, Accession # NP_722548
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4179
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.