Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD46 Antibody (MEM-258), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB500301APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500301APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500301APC Supplier Novus Biologicals Supplier No. NB500301APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD46 Monoclonal specifically detects CD46 in Human, Bovine samples. It is validated for Flow Cytometry.

Specifications

Antigen CD46
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-258
Conjugate APC
Dilution Flow Cytometry
Gene Alias AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen
Gene Symbols CD46
Host Species Mouse
Immunogen HPB-ALL human T cell line
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4179
Test Specificity The antibody MEM-258 recognizes an epitope on SCR4 (the membrane-proximal SCR) domain of CD46 (Membrane cofactor protein). CD46 is 56-66 kDa dimeric transmembrane protein expressed on T and B lymphocytes, platelets, monocytes, granulocytes, endothelial cells, epithelial cells and fibroblast; it is negative on erythrocytes.
Target Species Human, Bovine
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.