Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD46 Rabbit anti-Human, Clone: 1, Novus Biologicals™
SDP

Catalog No. NB021623 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB021623 100 μg
NB021624 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB021623 Supplier Novus Biologicals Supplier No. NBP290080100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD46 Monoclonal specifically detects CD46 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen CD46
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:100-1:500
Formulation 0.2 um filtered solution in PBS
Gene Alias AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen
Host Species Rabbit
Immunogen Recombinant Human CD46 protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4179
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.