Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD47 Antibody (CD47/3019), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB121118 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB121118 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB121118 Supplier Novus Biologicals Supplier No. NBP308976AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CD47
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CD47/3019
Conjugate Alexa Fluor 488
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen
Host Species Mouse
Immunogen A recombinant fragment (around aa 18-135) of human CD47 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 961
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.