Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD47 Antibody (IAP/964 + B6H12.2), HRP, Novus Biologicals™
SDP

Catalog No. NBP247812H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247812H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP247812H Supplier Novus Biologicals Supplier No. NBP247812H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD47 Monoclonal specifically detects CD47 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen CD47
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone IAP/964 + B6H12.2
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation PBS with No Preservative
Gene Alias antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen
Gene Symbols CD47
Host Species Mouse
Immunogen Recombinant human CD47 protein (CD47/964); Intact CD47 Purified from placenta (B6H12.2) (Uniprot: Q08722)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 961
Test Specificity This antibody reacts with Ig domain of CD47 protein. CD47, originally named integrin-associated protein (IAP), is a 50kDa protein containing five membrane-spanning sequences and a short cytoplasmic tail. CD47 plays a role in both cell adhesion by acting as an adhesion receptor for THBS1 on platelets, and in the modulation of integrins. It is important in memory formation and synaptic plasticity in the hippocampus. CD47 may play a role in membrane transport and/or integrin dependent signal transduction.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.