Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD47 Antibody (OX-101) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017788 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017788 0.025 mg
NB10066632 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017788 Supplier Novus Biologicals Supplier No. NB100666320.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD47 Monoclonal specifically detects CD47 in Rat samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.

Specifications

Antigen CD47
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone OX-101
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500
Formulation PBS with 0.09% Sodium Azide
Gene Accession No. P97829
Gene Alias antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen
Gene Symbols CD47
Host Species Mouse
Immunogen Rat thymocytes
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 961
Test Specificity Antibody recognizes the rat homologue of human CD47, which has been identified as the ligand for the rat signal regulatory protein (SIRP). Rat CD47 is expressed primarily by lymphocytes and macrophages but has also been detected at high levels on brain endothelial cells. Clone OX-101 has been reported to block the interaction of CD47 and SIRP.
Target Species Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.