Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD47 Antibody (OX-101) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | CD47 |
|---|---|
| Clone | OX-101 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot, Immunoprecipitation, Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017788
![]() |
Novus Biologicals
NB100666320.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10066632
![]() |
Novus Biologicals
NB10066632 |
0.125 mg |
Each for $499.50
|
|
|||||
Description
CD47 Monoclonal specifically detects CD47 in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.Specifications
| CD47 | |
| 1.0 mg/mL | |
| Western Blot, Immunoprecipitation, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Immunology, Signal Transduction | |
| P97829 | |
| 961 | |
| Rat thymocytes | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| OX-101 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Rat | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| CD47 | |
| IgG1 | |
| Protein G purified | |
| Antibody recognizes the rat homologue of human CD47, which has been identified as the ligand for the rat signal regulatory protein (SIRP). Rat CD47 is expressed primarily by lymphocytes and macrophages but has also been detected at high levels on brain endothelial cells. Clone OX-101 has been reported to block the interaction of CD47 and SIRP. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title