Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD48/SLAMF2 Antibody (007), Alexa Fluor™ 700
SDP

Catalog No. NB067508 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB067508 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB067508 Supplier Novus Biologicals Supplier No. NBP289640AF700

Rabbit Monoclonal Antibody

CD48/SLAMF2 Monoclonal specifically detects CD48/SLAMF2 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen CD48/SLAMF2
Applications ELISA
Classification Monoclonal
Clone 7
Conjugate Alexa Fluor 700
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD48/SLAMF2 (NP_001769.2; Met 1-Ser 220).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 962
Test Specificity No cross-reactivity in ELISA with: Mouse CD48 Human TNFRSF4
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.