Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD48/SLAMF2 Antibody (04), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB293220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB293220 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB293220 Supplier Novus Biologicals Supplier No. NBP326086C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD48/SLAMF2 Monoclonal antibody specifically detects CD48/SLAMF2 in Human samples. It is validated for ELISA

Specifications

Antigen CD48/SLAMF2
Applications ELISA
Classification Monoclonal
Clone 04
Conjugate DyLight 650
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2
Host Species Mouse
Immunogen This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 962
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.