Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD48/SLAMF2 Rabbit anti-Human, Clone: 248, Novus Biologicals™
SDP

Catalog No. NB020766 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020766 50 μg
NB020765 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020766 Supplier Novus Biologicals Supplier No. NBP28964150UL

Rabbit Monoclonal Antibody

CD48/SLAMF2 Monoclonal specifically detects CD48/SLAMF2 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD48/SLAMF2
Applications Flow Cytometry
Classification Monoclonal
Clone 248
Conjugate Unconjugated
Dilution Flow Cytometry 1:25-1:100
Formulation 0.2 um filtered solution in PBS
Gene Alias BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2
Host Species Rabbit
Immunogen Recombinant Human CD48/SLAMF2 protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 962
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.