Learn More
CD49d Mouse anti-Human, PE, Clone: 9F10, BD
Mouse Monoclonal Antibody
$116.50 - $279.50
Specifications
Antigen | CD49d |
---|---|
Clone | 9F10 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB560972
|
BD Biosciences
560972 |
25 Tests |
Each for $116.50
|
|
|||||
BDB555503
|
BD Biosciences
555503 |
100 Tests |
Each for $279.50
|
|
|||||
BDB556635
|
BD Biosciences
556635 |
50 Tests | N/A | N/A | N/A | ||||
Description
The 9F10 monoclonal antibody specifically reacts with the integrin α4 chain, that is expressed as a heterodimer with either of two β integrin subunits, β1 (CD29) or β7. The α4β1 integrin (VLA-4) is expressed on lymphocytes, monocytes, thymocytes, NK cells, and several B- and T-cell lines, and mediates binding to VCAM-1 (CD106) and the CS-1 region of fibronectin. The αβ7 also binds fibronectin and VCAM-1, and has been shown in the mouse to preferentially bind the mucosal vascular addressin molecule, MAdCAM-1. This antibody is useful for studies of the expression by and function of cells that express α4 chain-containing integrins.
Specifications
CD49d | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
9F10 | |
Monoclonal | |
Mouse | |
Baboon, Bovine, Canine, Cynomolgus, Equine, Feline, Human, Rhesus Monkey, Sheep | |
IgG1 κ | |
Affinity Purified | |
Integrin α4 chain; Integrin alpha 4; ITGA4; IA4; alpha 4 subunit of VLA-4 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.