Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD49f (Integrin alpha 6) Monoclonal Antibody (eBioGoH3 (GoH3)), Alexa Fluor™ 700, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
$435.00
Specifications
| Antigen | CD49f (Integrin alpha 6) |
|---|---|
| Clone | eBioGoH3 (GoH3) |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
56-049-582
![]()
|
Invitrogen™
56049582 |
100 μg |
Each for $435.00
|
|
|||||
56-049-580
|
Invitrogen™
56049580 |
25 μg | N/A | N/A | N/A | ||||
Description
CDf (Integrin alpha-6, ITGA6) is the integrin alpha chain alpha 6 protein. Integrins are integral cell-surface proteins composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. For example, alpha 6 may combine with beta 4 in the integrin referred to as TSP180, or with beta 1 in the integrin VLA-6. Integrins are known to participate in cell adhesion as well as cell-surface mediated signaling. Two transcript variants encoding different isoforms have been found for CDf. Functionally, integrins are cell surface receptors that interact with the extracellular matrix and mediate intracellular signals that affect cellular shape, mobility, and progression through the cell cycle in response to the extracellular matrix. Integrin receptors are composed of alpha and beta subunits, and form structural and functional linkages between the ECM (extra-cellular matrix) and intracellular cytoskeletal linker proteins. Signaling mediated by Intergrin/ECM interactions are also integrated with cellular responses to growth factor signaling to regulate cellular proliferation, cytoskeletal reorganization and other responses necessary for cellular survival. CDf is predominantly expressed by epithelia, and plays a critical structural role in the hemidesmosome. More than 18 alpha and 8 beta subunits with numerous splice variant isoforms have been identified in mammals. Certain integrins can also bind to soluble ligands or to counter-receptors on adjacent cells, such as the intracellular adhesion molecules (ICAMs), resulting in aggregation of cells.Specifications
| CD49f (Integrin alpha 6) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 5033401O05Rik; AI115430; Alpha6p; CD49 antigen-like family member F; CD49f; DKFZp686J01244; FLJ18737; integrin alpha 6; integrin alpha-6; Integrin alpha-6 heavy chain; Integrin alpha-6 light chain; integrin alpha6B; integrin subunit alpha 6; integrin, alpha 6; Itga6; ITGA6B; LOW QUALITY PROTEIN: integrin alpha-6; Processed integrin alpha-6; VLA-6 | |
| Itga6 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Itga6 |
| eBioGoH3 (GoH3) | |
| Flow Cytometry | |
| Alexa Fluor 700 | |
| Rat | |
| Human, Mouse | |
| P23229, Q61739 | |
| 16403, 3655 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title