Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5 Antibody (53-7.3), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB174913 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174913 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174913 Supplier Novus Biologicals Supplier No. FAB115K
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD5 Monoclonal specifically detects CD5 in Mouse samples. It is validated for Blocking B Cell Proliferation, Complement-dependent Cytotoxicity, CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen CD5
Applications Blocking Assay, Complement-dependent Cytotoxicity, CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 53-7.3
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Host Species Rat
Immunogen Mouse thymus or spleen
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 921
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.