Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5 Antibody (7.8) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB188097 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188097 0.1 mg
NB188120 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188097 Supplier Novus Biologicals Supplier No. NBP314758
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD5 Monoclonal antibody specifically detects CD5 in Human, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow (Cell Surface)

Specifications

Antigen CD5
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7.8
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 to 2 μg/mL, Flow Cytometry, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, Flow (Cell Surface)
Formulation PBS with 0.02% Sodium Azide
Gene Alias CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Host Species Mouse
Immunogen Human activated PBMC
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 921
Target Species Human, Mouse (Negative)
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2 α
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.