Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD5 Antibody (7.8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$201.00 - $310.00
Specifications
Antigen | CD5 |
---|---|
Clone | 7.8 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, Flow (Cell Surface) |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Description
CD5 Monoclonal antibody specifically detects CD5 in Human, Mouse (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Flow (Cell Surface)Specifications
CD5 | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology | |
PBS with 0.02% Sodium Azide | |
921 | |
IgG2 α | |
Protein A or G purified |
7.8 | |
Western Blot 1 to 2 μg/mL, Flow Cytometry, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, Flow (Cell Surface) | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse (Negative) | |
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1 | |
Human activated PBMC | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title