Learn More
CD5 Antibody (A25-G), Novus Biologicals™
SDP
Rabbit Monoclonal Antibody
Supplier:  Novus Biologicals NBP178994200UL
 View more versions of this product
Catalog No. NBP17899420
Only null left
Add to Cart
Description
Description
CD5 Monoclonal specifically detects CD5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CD5
Monoclonal
Unconjugated
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
CD5
Peptide derived from the C-terminal sequence of human CD10.
200 μL
Primary
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
A25-G
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Rabbit
Affinity Purified
RUO
921
Store at 4C. Do not freeze.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only