Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5 Antibody (A25-G), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$558.00 - $646.00

Specifications

Antigen CD5
Clone A25-G
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17899410
SDP
View Documents
Novus Biologicals
NBP178994100UL
100 μL
Each for $558.00
Only null left
Add to Cart
 
NBP17899420
SDP
View Documents
Novus Biologicals
NBP178994200UL
200 μL
Each for $646.00
Only null left
Add to Cart
 
Description

Description

CD5 Monoclonal specifically detects CD5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CD5
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200
Monoclonal
Rabbit
Human
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
CD5
Primary
Store at 4C. Do not freeze.
A25-G
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
20mM Tris/HCl (pH 8.0) and 20 mg/ml BSA with 0.05% Sodium Azide
921
Peptide derived from the C-terminal sequence of human CD10.
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.