Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5 Antibody (CRIS-1), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N234598PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N234598PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N234598PEC7 Supplier Novus Biologicals Supplier No. NBP234598PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD5 Monoclonal specifically detects CD5 in Human, Rhesus Macaque samples. It is validated for Flow Cytometry.

Specifications

Antigen CD5
Applications Flow Cytometry
Classification Monoclonal
Clone CRIS-1
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Gene Symbols CD5
Host Species Mouse
Immunogen Stimulated human leukocytes
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cytokine Research, Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 921.0
Test Specificity Recognizes a 67kDa transmembrane protein, which is identified as CD5 (HLDA I; WS Code T 29HLDA III; WS Code T 530). The CD5 antigen is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. Anti-CD5 is a pan T-cell marker that also reacts with a range of neoplastic B-cells, e.g. chronic lymphocytic leukemia/small lymphocytic lymphoma (CLL/SLL), mantle cell lymphoma, and a subset (∽10%) of diffuse large B-cell lymphoma. CD5 aberrant expression is useful in making a diagnosis of mature T-cell neoplasms.
Target Species Human, Primate
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.