Learn More
CD52 Antibody (Campath-1H) - Chimeric - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | CD52 |
| Applications | CyTOF, Dual RNAScope ISH-IHC, ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In vivo Assay, Western Blot |
| Classification | Monoclonal |
| Clone | Campath-1H |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Flow Cytometry 1:10 - 1:1000, ELISA, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay, CyTOF-ready |
| Formulation | PBS with 0.02% Proclin 300. with 0.02% Proclin 300 |
| Gene Alias | Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 |
| Gene Symbols | CD52 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.