Learn More
Description
Specifications
Specifications
| Antigen | CD52 |
| Applications | ELISA, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | Campath-1H |
| Conjugate | DyLight 405 |
| Dilution | ELISA, Immunohistochemistry, Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5 |
| Gene Symbols | CD52 |
| Host Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
