Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD52 Antibody (CD52/2276R), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB379337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB379337 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB379337 Supplier Novus Biologicals Supplier No. NBP308642V
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD52 Monoclonal antibody specifically detects CD52 in Human, Monkey samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CD52
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CD52/2276R
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5
Host Species Rabbit
Immunogen Recombinant full-length human CD52 protein (Uniprot: P31358)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1043
Target Species Human, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.