Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD52 Antibody (YTH 34.5-G2b (Campath-1G)), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB275899APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB275899APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB275899APC Supplier Novus Biologicals Supplier No. NBP275899APC
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD52 Monoclonal specifically detects CD52 in Human, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry.

Specifications

Antigen CD52
Applications Flow Cytometry
Classification Monoclonal
Clone YTH 34.5-G2b (Campath-1G)
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Cambridge pathology 1 antigen, CAMPATH-1 antigen, CD52 antigen, CD52 antigen (CAMPATH-1 antigen), CD52 molecule, CDw52, CDW52 antigen (CAMPATH-1 antigen), Epididymal secretory protein E5, HE5, Human epididymis-specific protein 5
Gene Symbols CD52
Host Species Rat
Immunogen Human peripheral mononuclear cells enriched for T-cells.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1043.0
Test Specificity Human CD52 antigen, also known as CAMPATH-1. CD52 is expressed at high density by lymphocytes, monocytes, eosinophils, thymocytes and macrophages. It is expressed by most lymphoid derived malignancies, although expression on myeloma cells is variable.
Target Species Human, Cynomolgus Monkey, Rhesus Macaque
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.