Learn More
CD52 Monoclonal Antibody (CF1D12), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$188.00 - $476.00
Specifications
Antigen | CD52 |
---|---|
Clone | CF1D12 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
63-052-941
![]() |
Invitrogen™
63052941 |
25 Tests |
Each for $188.00
|
|
|||||
63-052-942
![]() |
Invitrogen™
63052942 |
100 Tests |
Each for $476.00
|
|
|||||
Description
This CF1D12 monoclonal antibody reacts with human CD52, a cell surface glycoprotein expressed in the immune and reproductive systems. CF1D12 is thought to recognize the carbohydrate region of CD52, not the protein core of the molecule. Tissue-specific mod
CD52, also known as CAMPATH-1 antigen, is a glycoprotein that is expressed on the surface of various immune cells, including T and B lymphocytes, natural killer cells, and monocytes. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including multiple sclerosis, chronic lymphocytic leukemia, and autoimmune disorders. CD52 is the target of alemtuzumab, a monoclonal antibody that is used to treat these conditions by depleting immune cells that express CD52.Specifications
CD52 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
AI463198; B7; B7-Ag; cambridge pathology 1 antigen; CAMPATH-1; CAMPATH-1 antigen; Cd52; CD52 antigen; CD52 antigen (CAMPATH-1 antigen); CD52 molecule; CDW52; CDW52 antigen (CAMPATH-1 antigen); CLS1; Epididymal secretory protein E5; epididymis secretory sperm binding protein Li 171mP; he5; HEL-S-171mP; human epididymis-specific protein 5; Lymphocyte differentiation antigen B7; Mb7 | |
CD52 | |
IgG3 κ | |
Affinity chromatography | |
Antibody |
CF1D12 | |
Flow Cytometry | |
Super Bright 600 | |
Mouse | |
Human | |
P31358 | |
1043 | |
Human CD52. | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
CD52 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.