Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD52 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. p-7209626 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
Catalog No. Quantity
50-173-3597 150 μL
1 options

Catalog No. 50-173-3597

Supplier: Proteintech Group Inc 218091AP150UL

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD52 is a glycosylphosphatidylinositol (GPI) anchored antigen expression in a number of different tissues.

Specifications

Antigen CD52
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CD52
Gene Accession No. P31358
Gene Alias Cambridge pathology 1 antigen, CAMPATH 1 antigen, CD52, CD52 molecule, CDW52, He5
Gene Symbols CD52
Host Species Rabbit
Immunogen CD52 Fusion Protein Ag17788
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1043
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.