Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD53 Antibody (104), HRP, Novus Biologicals™
SDP

Catalog No. NB087526 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087526 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087526 Supplier Novus Biologicals Supplier No. NBP290952H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD53 Monoclonal antibody specifically detects CD53 in Cynomolgus Monkey samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CD53
Applications Western Blot, ELISA
Classification Monoclonal
Clone 104
Conjugate HRP
Formulation PBS
Gene Alias antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Cynomolgus CD53 (G7NW46; Glu107-Asn181).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 963
Target Species Cynomolgus Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.