Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD53 Antibody (MEM-53), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB50393A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50393A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50393A750 Supplier Novus Biologicals Supplier No. NB500393AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD53 Monoclonal specifically detects CD53 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen CD53
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MEM-53
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53
Gene Symbols CD53
Host Species Mouse
Immunogen Leukocytes of patient suffering from a LGL-type leukemia.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 963
Test Specificity The antibody MEM-53 reacts with CD53, a 32-40 kDa tetraspanin family glycoprotein exclusivelly expressed on leukocytes; it is not present on platelets, red blood cells and non-hematopoietic cells. The antibody MEM-53 reacts also with deglycosylated molecule (molecular weight of the antigen is reduced by 15 kDa using endoglycosidase F). HLDA IV; WS Code NL 59 HLDA V; WS Code B CD53.5 HLDA V; WS Code BP BP287 HLDA V; WS Code T T-096 HLDA V; WS Code X XB004
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.