Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD53 Mouse anti-Human, Clone: 161-2 (53/2), Novus Biologicals™
SDP

Catalog No. NB154524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB154524 0.025 mg
NB154525 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB154524 Supplier Novus Biologicals Supplier No. NBP3120760.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD53 Monoclonal antibody specifically detects CD53 in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen CD53
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 161-2 (53/2)
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias antigen MOX44 identified by monoclonal MRC-OX44, CD53 antigentetraspanin-25, CD53 glycoprotein, CD53 molecule, CD53 tetraspan antigen, cell surface antigen, Cell surface glycoprotein CD53, MOX44transmembrane glycoprotein, Tetraspanin-25, tspan-25, TSPAN25leukocyte surface antigen CD53
Host Species Mouse
Immunogen This antibody was raised by immunizng mice with stimulated human leukocytes.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 963
Target Species Human
Content And Storage Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.