Learn More
CD54 Mouse anti-Human, PE, Clone: HA58, BD
Mouse Monoclonal Antibody
$116.00 - $306.00
Specifications
Antigen | CD54 |
---|---|
Clone | HA58 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
The HA58 monoclonal antibody specifically binds to CD54 which is also known as ICAM-1 (intracellular adhesion molecule-1). CD54 is an 85-110 kDa type I transmembrane glycoprotein that belongs to the immunoglobulin supergene family. A soluble form of CD54 can also be found in biological fluids. CD54 is expressed on endothelial cells and both resting (weak) and activated (moderate) lymphocytes and monocytes. CD54 is a ligand for LFA-1 (CD11a/CD18) and Mac-1 (CD11b/CD18). The CD54 adhesion molecule plays roles in inflammatory and immune responses and neoplasia. This antibody (NA/LE format) blocks the mixed-lymphocyte reaction (MLR) and the purified format is suitable for staining acetone-fixed, frozen tissue sections.
Specifications
CD54 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
ICAM-1; intercellular adhesion molecule 1; BB2; P3.58 |
HA58 | |
Monoclonal | |
Mouse | |
Human | |
IFN-γ-treated BM314 colonic carcinoma cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.