Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD55/DAF Antibody (583905), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB368117 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368117 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368117 Supplier Novus Biologicals Supplier No. FAB5376K
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD55/DAF Monoclonal antibody specifically detects CD55/DAF in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen CD55/DAF
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 583905
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD55/DAF, Met1-Pro359, Accession # Q61475
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1604
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.