Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD55/DAF Antibody (67), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP250226UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250226UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250226UV Supplier Novus Biologicals Supplier No. NBP250226UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD55/DAF Monoclonal specifically detects CD55/DAF in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen CD55/DAF
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 67
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC
Gene Symbols CD55
Host Species Mouse
Immunogen Fibronectin purified human monocytes .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1604
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.