Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD55/DAF Antibody (BRIC216), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB065914549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB065914549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB065914549 Supplier Novus Biologicals Supplier No. NB10065914JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD55/DAF Monoclonal specifically detects CD55/DAF in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Western Blot.

Specifications

Antigen CD55/DAF
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BRIC216
Conjugate Janelia Fluor 549
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD55 antigen, CD55 molecule, decay accelerating factor for complement (Cromer blood group), CRdecay accelerating factor for complement (CD55, Cromer blood group system), CROMDAFcomplement decay-accelerating factor, decay accelerating factor for complement, TC
Gene Symbols CD55
Host Species Mouse
Immunogen Human fibroblast cell line
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 1604
Test Specificity NB100-65914 recognizes the CD55 antigen. BRIC 216 recognizes the consensus region 3 of the DAF molecule, which contains the functional site, and the antibody blocks the function of DAF.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.