Learn More
CD56 (NCAM-1) Mouse, anti-Human, R718, Clone: B159, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 566965
Description
The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.
Specifications
| CD56 (NCAM-1) | |
| Monoclonal | |
| R718 | |
| 100 Tests | |
| Primary | |
| Refrigerate Upon Arrival |
| Flow Cytometry | |
| B159 | |
| Mouse | |
| RUO | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.