Learn More
CD56 (NCAM-1) Mouse, anti-Human, R718, Clone: B159, BD Horizon™
Mouse Monoclonal Antibody
$144.60 - $303.50
Specifications
Antigen | CD56 (NCAM-1) |
---|---|
Clone | B159 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Description
The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.
Specifications
CD56 (NCAM-1) | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
Refrigerate Upon Arrival |
B159 | |
Monoclonal | |
Liquid | |
RUO | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.