Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD57 Antibody (SPM129) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP247790B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247790B 0.2 mg
NBP247790A 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP247790B Supplier Novus Biologicals Supplier No. NBP2477900.2MG

Mouse Monoclonal Antibody

beta-1,3-Glucuronyltransferase 1/B3GAT1 Monoclonal antibody specifically detects Antigen in Human, Rat (Negative) samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen beta-1,3-Glucuronyltransferase 1/B3GAT1
Applications Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone SPM129
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 2 - 4 ug/ml, Immunofluorescence : 0.5 1 ug/ml
Formulation PBS with No Preservative
Gene Alias Beta-1,3-glucuronyltransferase 1, beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P), EC 2.4.1.135, galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, GlcAT-P, GLCATPGLCUATP, glcUAT-P, Glucuronosyltransferase P, NK1, NK-1, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
Gene Symbols B3GAT1
Host Species Mouse
Immunogen Human peripheral blood mononuclear cells
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 27087
Test Specificity Anti-CD57 marks a subset of lymphocytes known as natural killer (NK) cells. Follicular center cell lymphomas often contain many NK cells within the neoplastic follicles. Anti-CD57 also stains neuroendocrine cells and their derived tumors, including carcinoid tumor and medulloblastoma. Anti-CD57 can also be useful in separating type B3 thymoma from thymic carcinoma when combined with a panel that includes antibodies against GLUT1, CD5, and CEA.
Target Species Human, Rat (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.