Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD58/LFA-3 Antibody (185), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB037375 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB037375 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB037375 Supplier Novus Biologicals Supplier No. NBP290098G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD58/LFA-3 Monoclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot.

Specifications

Antigen CD58/LFA-3
Applications Western Blot
Classification Monoclonal
Clone 185
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CD58/LFA-3 (AAH05930.1; Met1-Arg215).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 965
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.