Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD58/LFA-3 Antibody (MM0176-8G24), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N212172APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N212172APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N212172APC7 Supplier Novus Biologicals Supplier No. NBP212172APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD58/LFA-3 Monoclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CD58/LFA-3
Applications Flow Cytometry
Classification Monoclonal
Clone MM0176-8G24
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3
Gene Symbols CD58
Host Species Mouse
Immunogen Human recombinant CD58/LFA-3 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cell Biology, Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 965.0
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.