Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD58/LFA-3 Antibody (TS2/9) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB437644 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
50 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB437644 50 ug
NBP222542 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB437644 Supplier Novus Biologicals Supplier No. NBP22254250UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD58/LFA-3 Monoclonal specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready, Block/Neutralize.

Specifications

Antigen CD58/LFA-3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF, Neutralization
Classification Monoclonal
Clone TS2/9
Concentration 0.98 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:500, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, CyTOF-ready, Neutralization
Gene Accession No. P19256
Gene Alias Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3
Gene Symbols CD58
Host Species Mouse
Immunogen Human Leukocyte Function Antigen-3
Purification Method Protein A purified
Quantity 50 ug
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 965
Test Specificity This antibody reacts with Human.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.