Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD59 Antibody (28D4), Novus Biologicals™
SDP

Catalog No. NB297625 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297625 100 μL
NB297624 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297625 Supplier Novus Biologicals Supplier No. NBP326676100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD59 Monoclonal antibody specifically detects CD59 in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen CD59
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 28D4
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias 16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein
Host Species Rabbit
Immunogen A synthesized peptide derived from Human CD59 [UniProt P13987]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 966
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.