Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD59 Rabbit anti-Rat, Clone: 2, Novus Biologicals™
SDP

Catalog No. NB025629 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB025629 50 μg
NB025628 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB025629 Supplier Novus Biologicals Supplier No. NBP29091050UL

Rabbit Monoclonal Antibody

CD59 Monoclonal specifically detects CD59 in Rat samples. It is validated for Sandwich ELISA Capture.

Specifications

Antigen CD59
Applications ELISA
Classification Monoclonal
Clone 2
Conjugate Unconjugated
Dilution Sandwich ELISA Capture 1:250-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias 16.3A5, 1F5, 1F5 antigen, 20 kDa homologous restriction factor, CD59 antigen, CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, CD59 antigen, complement regulatory protein, CD59 glycoprotein, CD59 molecule, complement regulatory protein, EJ16, EJ30, EJ30, EL32 and G344), EL32, FLJ38134, FLJ92039, G344, HRF20, HRF-20, human leukocyte antigen MIC11, Ly-6-like protein, lymphocytic antigen CD59/MEM43, MACIF, MAC-inhibitory protein, MAC-IP, MEM43, MEM43 antigen, membrane attack complex (MAC) inhibition factor, Membrane attack complex inhibition factor, Membrane inhibitor of reactive lysis, MGC2354, MIC11MSK21, MIN1, MIN2, MIN3, MIRL, p18-20, protectin, surface anitgen recognized by monoclonal 16.3A5, T cell-activating protein
Host Species Rabbit
Immunogen Recombinant Rat CD59 Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Immunology, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 966
Target Species Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.