Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD5L Antibody (38), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB072518 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072518 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072518 Supplier Novus Biologicals Supplier No. NBP290356AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD5L Monoclonal specifically detects CD5L in Mouse samples. It is validated for ELISA.

Specifications

Antigen CD5L
Applications ELISA
Classification Monoclonal
Clone 38
Conjugate Alexa Fluor 647
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias AIM, API6, apoptosis inhibitor 6, CD5 antigen-like, CD5 antigen-like (scavenger receptor cysteine rich family), CD5 molecule-like, CT-2, IgM-associated peptide, PRO229, Spalpha, SP-ALPHA
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD5L (Met 1-Val 352; NP_033820.2) .
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 922
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.