Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD6 Antibody (001), Alexa Fluor™ 488
SDP

Catalog No. NB040649 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB040649 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB040649 Supplier Novus Biologicals Supplier No. NBP290690AF488

Rabbit Monoclonal Antibody

CD6 Monoclonal specifically detects CD6 in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CD6
Applications Immunofluorescence
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 488
Dilution Immunocytochemistry/Immunofluorescence, Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD6 (Q91WN5; Met1-Gly396).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 923
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.